Learn More
Sensidyne™ Detector Tubes: Water Vapor
SDP
Catalog No. 18074J Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
0.05 to 2mg/L
Catalog No. Measurement Range
18-074J 0.05 to 2mg/L
1 options
Catalog No. 18-074J Supplier Sensidyne™ Supplier No. 177U
All detector tubes are sold in packs of 10.
Specifications
Components Measured Water Vapor
Measurement Range 0.05 to 2mg/L
No. of Strokes 1
Quantity 10
Response Time 1 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C