Learn More
Meridian Life Science SENSIFAST PROBE HI-ROX ONE-ST
SDP
Supplier:  Meridian Life Science BIO77005
Encompass
SensiFAST Probe Hi-ROX One-Step Kit has been specially formulated for highly reproducible first-strand cDNA synthesis and subsequent real-time PCR in a single tube.The kit is designed for use with probe-detection technology.
Catalog No. NC0595765
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.