Learn More
Vernier Sensor Bracket for Go Direct™
SDPSCI_ED
Catalog No. S1724324 Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Go Direct™ sensors
Catalog No. For Use With (Equipment)
S1724324 Go Direct™ sensors
1 options
Catalog No. S1724324 Supplier Vernier Supplier No. GDXMOUNT
Add to Cart
Add to Cart
This bracket fits most of Vernier's box-shaped Go Direct™ sensors. Attach the bracket to Vernier's accessory rod or any tripod with a standard camera mount to hold a sensor still and stable for repetitive measurements.
This accessory allows mounting of the following sensors:
  • Go Direct™ Sound Sensor
  • Go Direct™ Gas Pressure Sensor
  • Go Direct™ Light and Color Sensor
  • Go Direct™ Radiation Monitor
  • Go Direct™ EKG Sensor
  • Go Direct™ Energy Sensor
  • Go Direct™ Current Probe
  • Go Direct™ Voltage Probe
  • Go Direct™ Constant Current System
The bracket fits on these items:
  • Replacement Rod for Dual-Range Force Sensor, Photogate, or WDSS
  • Light Sensor Holder for Optics Expansion Kit
Specifications
Description Threaded insert: 0.25 in. diameter, 20 threads/in.
Includes 1 sensor bracket
Material Plastic
For Use With (Application) Used to mount a Go Direct™ sensor to a photo tripod
For Use With (Equipment) Go Direct™ sensors