Learn More
Cytiva SENSOR CHIP CM3 PACK OF 3
Supplier:  Cytiva BR100541
Encompass
An alternative to Sensor Chip CM5 with shorter dextran matrix and similar charge density - to explore alternative assay conditions.
Catalog No. 50-105-5325