Learn More
Cytiva SENSOR CHIP NTA, (3-PACK)
Catalog No. 501055277 Shop All Cytiva Products
Encompass
Sensor chip NTA, used for convenient capture of histidine-tagged molecules.
Catalog No. 50-105-5277 Supplier Cytiva Supplier No. BR100034
Sensor chip NTA, used for convenient capture of histidine-tagged molecules.