Learn More
Cytiva SENSOR CHIP PROTEIN A 3-PACK
Supplier:  Cytiva 29127558
Encompass
Sensor Chip Protein A, Pack of 3, Ensures orientation-specific binding of antibodies through the Fc portion.
Catalog No. 50-105-5506