Learn More
Cytiva SENSOR CHIP CM5 (10/PK)
Supplier: Cytiva 29149604
Sensor Chip CM5, pack of 10, is the most versatile chip available the first choice for immobilization via -NH2, -SH, -CHO, -OH or -COOH groups.