Learn More
Cytiva SENSOR CHIP SA, (3-PACK)
Catalog No. 501055276 Shop All Cytiva Products
Encompass
Sensor Chip SA, is used for stable and convenient immobilization of biotinylated molecules.
Catalog No. 50-105-5276 Supplier Cytiva Supplier No. BR100032
Sensor Chip SA, is used for stable and convenient immobilization of biotinylated molecules.