Learn More
ChemDAQ™ Sensor Communication Interface Intrinsically Safe Oxygen Package - includes Monitor, Barrier Unit, and Remote Sensor Mount
Catalog No. 17987113
Change view
Click to view available options
For Use With (Equipment):
ChemDAQ Gas Sensor
Catalog No. For Use With (Equipment)
17-987-113 ChemDAQ Gas Sensor
1 options
Catalog No. 17-987-113 Supplier ChemDAQ™ Supplier No. SCIISO2PKG
Add to Cart
Add to Cart
Request a Quote
Sensor Communication Interface Intrinsically Safe Oxygen Package delivers continuous O2 monitoring in hazardous locations. Includes monitor, intrinsically safe barrier unit, and remote sensor mount for safe, accurate detection and integration with facility safety systems.
The Sensor Communication Interface Intrinsically Safe Oxygen Package is a comprehensive gas detection solution engineered to provide continuous, real-time monitoring of oxygen (O2) levels in hazardous or classified environments where explosion risks are present. Designed specifically to meet the stringent safety requirements of hazardous locations, this package ensures accurate oxygen measurement while maintaining compliance with intrinsic safety standards.
Key Features
  • Intrinsically Safe Design: The package includes an intrinsically safe barrier unit that limits electrical and thermal energy, preventing sparks or excessive heat that could ignite flammable atmospheres. This critical safety feature makes the system suitable for use in classified hazardous areas such as chemical plants, pharmaceutical facilities, and sterilization rooms.
  • Continuous Oxygen Monitoring: Provides real-time data on ambient oxygen levels, allowing early detection of oxygen deficiency or enrichment. This is essential for worker safety in confined spaces, cleanrooms, and controlled environments where oxygen displacement by inert gases or combustion can occur.
  • Remote Sensor Mount: The included remote sensor mount allows the oxygen sensor to be placed precisely at the point of potential risk, such as inside chambers, confined spaces, or close to chemical storage, while the monitor and electronics remain in a safe, accessible location.
  • Seamless Integration & Communication: Equipped with standard industrial communication protocols including Modbus RTU, 4–20 mA analog output, and dry contact relays. This interface module can easily connect to existing Building Management Systems (BMS), Programmable Logic Controllers (PLCs), alarm systems, and safety interlocks, supporting facility-wide safety monitoring and automation.
  • User-Friendly Interface & Configurable Alarms: The monitor provides a clear digital display of oxygen levels with configurable alarm thresholds and visual/audible alerts, enabling rapid response to unsafe conditions. This flexibility allows customization to meet site-specific safety standards and regulations.
  • Modular Sensor System (SXP™): Utilizes ChemDAQ's patented SXP™ sensor cartridges, which are pre-calibrated and easily replaceable without downtime or special tools. This design reduces maintenance complexity and ensures consistent, accurate sensor performance over time.
Oxygen levels in industrial and classified environments must be carefully controlled to ensure safety and maintain regulatory compliance. The Sensor Communication Interface Intrinsically Safe Oxygen Package offers a robust, dependable solution that combines precision sensing, flexible installation, and safety certifications to protect personnel and assets. Its intrinsically safe barrier unit guarantees safe operation in potentially explosive atmospheres, while the modular sensor system and remote mount provide adaptability for a wide range of applications. Whether monitoring oxygen displacement in inert gas environments or ensuring safe air quality in confined spaces, this package delivers peace of mind and operational reliability.
Specifications
For Use With (Equipment) ChemDAQ Gas Sensor
Type Sensor Communication Interface
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.