Learn More
ChemDAQ™ Sensor Communication Interface Module for Ethylene Oxide (Includes Remote Sensor Mount)
Catalog No. 17987109
Change view
Click to view available options
For Use With (Equipment):
ChemDAQ Gas Sensor
Catalog No. For Use With (Equipment)
17-987-109 ChemDAQ Gas Sensor
1 options
Catalog No. 17-987-109 Supplier ChemDAQ™ Supplier No. SCIESCIM
Add to Cart
Add to Cart
Sensor Communication Interface Module for Hydrogen Peroxide provides real-time H2O2 monitoring with seamless system integration. Designed for accurate detection and data output to facility controls. Ideal for sterilization areas and safety-critical environments.
The Sensor Communication Interface Module for Ethylene Oxide (EtO) provides continuous, real-time monitoring of ethylene oxide vapor levels to ensure safety and regulatory compliance in environments where EtO is present. Designed for integration with facility safety systems, this module offers reliable detection and flexible installation options.
Key Features
  • Continuous EtO Vapor Detection Monitors ethylene oxide vapor concentrations in real-time, providing early warning of leaks or unsafe conditions to protect personnel and equipment.
  • Remote Sensor Mount Included Allows the sensor to be placed directly at critical risk points, such as sterilizers or storage areas, while keeping the monitor in a safe, accessible location.
  • Versatile Communication Options Supports multiple output protocols, including:
    • Modbus RTU
    • 4–20 mA Analog Output
    • Dry Contact Relays
    This enables seamless integration with Building Management Systems (BMS), Programmable Logic Controllers (PLC), and alarm systems for automated safety responses.
  • Modular SXP™ Sensor Cartridge System Uses ChemDAQ's patented, pre-calibrated SXP™ electrochemical sensors, allowing easy replacement without specialized tools, reducing maintenance time and costs.
  • Configurable Alarms and User Interface Features a clear display with customizable visual and audible alarms for immediate notification of hazardous EtO levels.
This module is a robust solution for maintaining safe ethylene oxide environments, combining advanced sensor technology, intrinsic safety features, and flexible integration capabilities to meet your facility's monitoring needs.Purchase of this product requires and annual service contract for sensor calibration and exchange.
Specifications
For Use With (Equipment) ChemDAQ Gas Sensor
Type Sensor Communication Interface
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.