Learn More
ChemDAQ™ Sensor Communication Interface Module for Hydrogen Peroxide
Description
The Sensor Communication Interface Module for Hydrogen Peroxide is a precision monitoring solution designed to provide continuous, real-time detection of hydrogen peroxide (H2O2) vapor levels in a variety of industrial and sterilization environments. This module offers reliable, accurate measurements to ensure workplace safety, regulatory compliance, and process control.
Key Features
- Continuous Hydrogen Peroxide Monitoring This interface module continuously measures hydrogen peroxide vapor concentrations, allowing early detection of leaks or elevated levels. Real-time monitoring helps protect workers, prevent contamination, and maintain safe operating conditions.
- Flexible Communication and Integration The module supports multiple communication protocols to integrate seamlessly into existing safety and automation systems, including:
- Modbus RTU
- 4–20 mA Analog Output
- Dry Contact Relays
- User-Friendly Interface Clear digital display and configurable alarms provide visual and audible alerts, ensuring rapid response to unsafe hydrogen peroxide vapor levels.
- Modular SXP™ Sensor Cartridge System Utilizes ChemDAQ's proprietary SXP™ electrochemical sensor cartridges, which are factory-calibrated and designed for easy, tool-free replacement reducing downtime and maintenance costs.
- Remote Sensor Mount Capability Allows the sensor to be positioned in high-risk locations (e.g., near sterilizers or storage tanks), while the main module remains accessible and safe, enhancing detection accuracy and ease of use.
Hydrogen peroxide is a widely used sterilant but poses safety risks if vapor concentrations are not carefully monitored. The Sensor Communication Interface Module for Hydrogen Peroxide offers a reliable, easy-to-integrate solution to maintain safe environments in critical applications. Its advanced sensor technology, flexible communication capabilities, and modular design provide exceptional accuracy and ease of maintenance. Ideal for healthcare, pharmaceutical, and industrial settings, this module supports safety, compliance, and operational efficiency.
Specifications
Specifications
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Sensor Communication Interface |
Purchase of this product requires an annual service contract for sensor calibration and exchange, which must be arranged prior to shipment. Customers in the food and beverage production and processing industry need to contact ChemDAQ directly to purchase this product. Contact your Fisher Scientific representative to confirm your needs and to request a quote.