Learn More
ChemDAQ™ Sensor Communication Interface Module for Peracetic Acid
Catalog No. 17987114
Change view
Click to view available options
For Use With (Equipment):
ChemDAQ Gas Sensor
Catalog No. For Use With (Equipment)
17-987-114 ChemDAQ Gas Sensor
1 options
Catalog No. 17-987-114 Supplier ChemDAQ™ Supplier No. SCIPSCIM
Add to Cart
Add to Cart
Sensor Communication Interface Module for Peracetic Acid provides continuous, real-time monitoring of PAA vapors. Designed for seamless integration with facility systems, it ensures accurate detection and supports safety compliance in sterilization and industrial environments.
The Sensor Communication Interface Module for Peracetic Acid (PAA) is a specialized gas detection system designed to provide continuous, real-time monitoring of peracetic acid vapor concentrations in critical environments. Ideal for use in sterilization rooms, pharmaceutical manufacturing, cleanrooms, and other industrial settings where PAA is used as a disinfectant or sterilant, this interface module delivers precise, reliable detection to help maintain safe working conditions and regulatory compliance.
Key Features
  • Real-Time Peracetic Acid Vapor Detection The module continuously monitors airborne PAA levels, providing instantaneous readings as well as time-weighted average (TWA) values. This allows facility managers and safety personnel to promptly identify hazardous concentrations and take necessary action.
  • Seamless System Integration Designed to integrate smoothly with existing facility management and control systems, the module supports standard industrial communication protocols such as:
    • Modbus RTU
    • 4–20 mA Analog Output
    • Dry Contact Relays
    This ensures compatibility with building management systems (BMS), programmable logic controllers (PLCs), alarm systems, and abatement controls.
  • Remote Sensor Mount Capability For optimal detection accuracy, the module supports a remote sensor mount, allowing the sensor to be positioned close to potential PAA emission sources such as sterilizers, ventilation points, or storage areas.
  • Pre-Calibrated Sensor Exchange (SXP™) System The module uses ChemDAQ's innovative SXP™ sensor cartridges, which are pre-calibrated and easily replaceable without requiring specialized tools or downtime. This minimizes maintenance effort and keeps monitoring systems running smoothly.
  • Configurable Alarms and Indicators The interface module features customizable visual and audible alarms, providing immediate notification when PAA vapor concentrations exceed user-defined thresholds. These alerts help ensure worker safety and facilitate rapid response to hazardous conditions.
Peracetic acid is an effective sterilant widely used in healthcare, pharmaceutical, and industrial applications. However, exposure to elevated PAA vapors poses significant health risks. The Sensor Communication Interface Module for Peracetic Acid offers a reliable, user-friendly solution for continuous gas detection in demanding environments. Its advanced sensing technology, combined with flexible installation options and seamless system integration, makes it an essential component for any facility aiming to protect personnel, maintain safe air quality, and comply with strict safety standards.
Specifications
For Use With (Equipment) ChemDAQ Gas Sensor
Type Sensor Communication Interface
Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.