Learn More
Cytiva (Formerly Pall Lab) Sentino Filter Funnel, 100 mL, Gamma Irradiated, 25/sleeve (100/pkg)
Catalog No. 502063110 Shop All Cytiva Products
Encompass
Maximize Workspace and Minimize Contamination Risk-Small footprint, short height, light design and battery operation option mean its ideal anywhere on the bench or in the field.-Simple to load and disposable to eliminate potential biofilm build-up. There is no need to clean, wrap, and autoclave a bulky multi-place manifold. -Pulls sample through the filter and fluid path eliminating the need for a vacuum source and reducing the risk of back-up and contamination of analysis.-Meets the Membrane Filter (MF) Technique filtration parameters described in U.S. EPA, ISO, and ASTM standards. -Eliminates a fluid trap under the vacuum, preventing implosion accidents.-Shape of unit and materials of construction are easy to wipe down with standard laboratory disinfectants.-Soft-touch keypad has simple on/off function and pulse features to control filtration. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3110 Supplier Cytiva Supplier No. 4870
Maximize Workspace and Minimize Contamination Risk-Small footprint, short height, light design and battery operation option mean its ideal anywhere on the bench or in the field.-Simple to load and disposable to eliminate potential biofilm build-up. There is no need to clean, wrap, and autoclave a bulky multi-place manifold. -Pulls sample through the filter and fluid path eliminating the need for a vacuum source and reducing the risk of back-up and contamination of analysis.-Meets the Membrane Filter (MF) Technique filtration parameters described in U.S. EPA, ISO, and ASTM standards. -Eliminates a fluid trap under the vacuum, preventing implosion accidents.-Shape of unit and materials of construction are easy to wipe down with standard laboratory disinfectants.-Soft-touch keypad has simple on/off function and pulse features to control filtration. This product is not approved for use in medical, clinical, surgical or other patient applications.