Learn More
Kimberly-Clark™ Professional Sentry III 6pt Ratchet Hard Hat
Description
- Low-profile, silhouette-styled
- Meet ANSI Z89.1 - 2009 requirements for impact (as well as Class C, G and E)
- Six-point ratchet suspension system and a flat, customizable front area
- Customize to suit needs: Fits head sizes 6.5 to 8 and have accessory slots for attaching additional compatible personal protective equipment
Specifications
Specifications
| Color | Green |
| Size | 6.5 to 8 |