Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terra™ Sentry NT FP IMG ESR 6 in. Brown Boot

Safety boot built with a lower profile and grounded with an aggressive new outsole for improved, all season-slip resistance.

Supplier:  Terra™ R4NWBN 140 W

 View more versions of this product

Catalog No. 17-100-077


Only null left
Add to Cart

Description

Description

  • Full-grain waterproof sealed-seam leather upper
  • Four-row stitched combat upper pattern with comfort bellows tongue
  • Abrasion-resistant TECTUFF™ leather back strap
  • Moisture-wicking lining with CLEANFEET™ odor-control treatment
  • XRD™ Internal MET guard
  • All-season, slip-resistant, heavy-duty TPU outsole
  • Direct-injected, lightweight and shock-absorbing PU midsole, with PU toe bumper
  • Premium comfort PU FOOTBED™ insole
Specifications

Specifications

Brown
14W
No
Leather
Men's
Work Boot
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.