Learn More
Terra™ Sentry NT FP IMG ESR 6 in. Brown Boot
Catalog No. 17100088 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
10.5
Catalog No. Size
17-100-088 10.5
1 options
Catalog No. 17-100-088 Supplier Terra™ Supplier No. R4NWBN 105 M
Add to Cart
Add to Cart
Safety boot built with a lower profile and grounded with an aggressive new outsole for improved, all season-slip resistance.
  • Full-grain waterproof sealed-seam leather upper
  • Four-row stitched combat upper pattern with comfort bellows tongue
  • Abrasion-resistant TECTUFF™ leather back strap
  • Moisture-wicking lining with CLEANFEET™ odor-control treatment
  • XRD™ Internal MET guard
  • All-season, slip-resistant, heavy-duty TPU outsole
  • Direct-injected, lightweight and shock-absorbing PU midsole, with PU toe bumper
  • Premium comfort PU FOOTBED™ insole
Specifications
Color Brown
Material Leather
Size 10.5
Gender Men's
Latex Free Yes
Waterproof No
Product Type Work Boot