Learn More
Waters Corp Sep-Pak Florisil Classic Cartridge, 900 mg Sorbent per Cartridge, 50 - 200 µm, 50/pk
Supplier:  Waters Corp WAT051960
Encompass_Preferred
Polar, highly active, weakly basic sorbent for adsorption of low to moderate polarity species from nonaqueous solutions.
Catalog No. 50-785-658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.