Learn More
GVS Separa Toggler
Catalog No. 09316829 Shop All Osmonics Products
Change view
Click to view available options
For Use With (Equipment):
Syringe Filters
Catalog No. For Use With (Equipment)
09-316-829 Syringe Filters
1 options
Catalog No. 09-316-829 Supplier GVS Supplier No. SEPARATOGGLER120
Only null left
Add to Cart
Add to Cart
GVS Separa Toggler is a Multi Vial Compressor.
  • Further shorten 30% sample preparation time
  • Multiple filtration of 12 SEPARA™ syringeless filter vial samples at a time
  • Reduce user hand stress
Specifications
Type Multi-vial Compressor
For Use With (Equipment) Syringe Filters