Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Separase Monoclonal Antibody (XJ11-4D7)
GREENER_CHOICE

Catalog No. AMPA6573
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6573 Supplier Invitrogen™ Supplier No. MA116573
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Separase Monoclonal Antibody for ICC/IF

Caspase-like protease, which plays a central role in the chromosome segregation by cleaving the SCC1/RAD21 subunit of the cohesin complex at the onset of anaphase. During most of the cell cycle, it is inactivated by different mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Separase
Applications Immunocytochemistry
Classification Monoclonal
Clone XJ11-4D7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene ESPL1
Gene Accession No. P60330, Q14674
Gene Alias AL024103; AU045071; Caspase-like protein ESPL1; Cerp; Cut1/ESP1 related protein; ESP1; ESPL1; extra spindle pole bodies 1 (S. cerevisiae); extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle pole bodies homolog 1 (S. cerevisiae); extra spindle pole bodies like 1, separase; extra spindle poles like 1; extra spindle poles-like 1 protein; FLJ46492; KIAA0165; PRCE; SEPA; separase; Separin; separin, cysteine protease; SSE
Gene Symbols ESPL1
Host Species Mouse
Immunogen Maltose-Binding Protein fusion of C-terminal fragment of human Separase (amino acids 1866-1996).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105988, 9700
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.