Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Separase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139984 Supplier Invitrogen™ Supplier No. PA5139984
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TQKAAVETSF LDYGENLVQK WQVLSEVLSC SEKLVCHLGR LGSVSEAKAF CLEALKLTTK LQIPRQCALF LVLKGELELA RNDIDLCQSD LQQVLFLLES CTEFGGVTQH LDSVKKVHLQ KGKQQAQVPC PPQLPEEELF LRGPALELVA Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 85%.

Caspase-like protease, which plays a central role in the chromosome segregation by cleaving the SCC1/RAD21 subunit of the cohesin complex at the onset of anaphase. During most of the cell cycle, it is inactivated by different mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Separase
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ESPL1
Gene Accession No. Q14674
Gene Alias AL024103; AU045071; Caspase-like protein ESPL1; Cerp; Cut1/ESP1 related protein; ESP1; ESPL1; extra spindle pole bodies 1 (S. cerevisiae); extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle pole bodies homolog 1 (S. cerevisiae); extra spindle pole bodies like 1, separase; extra spindle poles like 1; extra spindle poles-like 1 protein; FLJ46492; KIAA0165; PRCE; SEPA; separase; Separin; separin, cysteine protease; SSE
Gene Symbols ESPL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ESPL1. Recombinant protein control fragment (Product #RP-109327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.