Learn More
CMP SCIENTIFIC CORP Separation capillary, PS1 coated
Supplier:  CMP SCIENTIFIC CORP ESCPS1360/3205065B1
Encompass
Separation capillary (1 per box): 360/320 µm OD, 50 µm ID, 65 cm, PS1 coated
Catalog No. NC2318824
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.