Learn More
EKF HemataSEAL™ Tube Sealant
Catalog No. 23550112 Shop All Separation Technology Products
Change view
Click to view available options
Quantity:
10 Pk.
Catalog No. Quantity
23-550-112 10 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-550-112 Supplier EKF Supplier No. 260100
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Clay-based sealant maintains excellent retention during centrifugation
  • Nontoxic, nonhardening, nondrying
  • Light color provides a sharp contrast line with blood for easy reading
  • Each plastic tray has 24 numbered positions for sample identification
  • One tray seals approximately one vial of tubes
  • Seals glass and plastic tubes
Order Info
Box of 10 trays
Specifications
Material Clay
Product Type Tube Sealant
For Use With (Application) Maintains excellent retention during centrifugation
For Use With (Equipment) Glass and Plastic tubes
No. of Places 24.0
Quantity 10 Pk.