Learn More
Sepax Technologies Inc Sepax HPLC Column Bioservice Affinity HSA, PEEK, 4.6 x 50 mm
SDP
Supplier:  Sepax Technologies Inc 272101P4605
Encompass_Preferred
Sepax offers an off-the-shelf product range of affinity columns conjugated with Human Serum Albumin (HSA) for our customers who are looking to asset mAb Immunogen Cross-Reactivity. The columns available can be easily attached to standard HPLC/UPLC instruments and provide an unique solution for process monitoring. For customers interested in custom immobilization contact us for more information.
Catalog No. 50-233-6951
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.