Learn More
Sepax Technologies Inc Sepax HPLC Column Monomix MC10-SEC, 10 µm, 500 Å, 7.8 x 300 mm
SDP
Supplier:  Sepax Technologies Inc 2801105007830
Encompass_Preferred
Monomix MC SEC bulk media are highly crosslinked spherical resins made of polymethylacrylate. These rigid resins are narrowly dispersed particles with particle size selection of 10 μm and pore size selection of 500 and 1000 Å. These resins have proprietary hydrophilic surface functional groups that minimize non-specific binding. Monomix MC SEC resins possess several key benefits: a broad pH tolerant range (1-14), elevated operating temperature (up to 80°C), high operating pressure (up to 20 bar), monosized particle (D90/D10 < 1.3, low column back pressure, high theoretical plate number), smooth surface and highly spherical (for easy column packing).
Catalog No. 50-233-8152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.