Learn More
Sepax Technologies Inc Sepax HPLC Column Unix™ SEC-300, 1.8 µm, 4.6 x 250 mm
SDP
Supplier:  Sepax Technologies Inc 2113004625
Encompass_Preferred
Utilizing proprietary surface technologies, Unix SEC phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 µm. The combination of small particle size and large pore volume of Unix SEC renders the highest separation efficiency and resolution of analytes. The well-controlled surface chemistry results in excellent lot-to-lot reproducibility. Our unique bonding chemistry, coupled with the maximized bonding density, allows Unix SEC to provide high stability and negligible non-specific interactions. Typical applications for Unix SEC columns include separation and analysis of biological molecules and water-soluble polymers.
Catalog No. 50-233-6725
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.