Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sephadex™ G-100 Media

Catalog No. 45000025 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
1 mL/min.
4.2 mL/min.
Particle Size:
100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Particle Size
45-000-025 4.2 mL/min. 100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
45-000-027 1 mL/min. 25 to 100 μm (Wet Form), 10 to 40 μm (Dry Form)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-025 Supplier Cytiva Supplier No. 17006001
Add to Cart
Add to Cart

Commonly used for desalting and buffer exchange of very large biomolecules

  • Available in two particle size ranges

Fine particle size

  • Particle size, dry: 40 to 120μm
  • Fractionation range, globular proteins: 4000 to 150,000
Superfine particle size
  • Particle size, dry: 10 to 40μm
  • Fractionation range, globular proteins: 4000 to 100,000

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Flow Rate 4.2 mL/min.
Particle Size 100 to 310 μm (Wet Form), 100 to 310 μm (Dry Form)
Bed Volume 15 to 20mL/g in Distilled Water
Product Type Sephadex G-100 Media
Format Solid Bead
For Use With (Application) Useful for desalting and Buffer exchange of very large biomolecules
Max. Pressure 0.096 bar
pH Range 2 to 10
Support Type Cross-linked Dextran
Sample Type Large biomolecules
Content And Storage 4° to 30°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.