Learn More
Rockland Immunochemicals SEPHAROSE™ PROTEIN A - PA50-00-0002
SDP
Supplier:  Rockland Immunochemicals PA50000002
Encompass_Preferred
Sepharose™ Protein A is a suspension of beads conjugated to native protein A derived from Staphylococcus aureaus. When evenly dispersed approximately 2.0 cc of suspension is equal to 1.0 cc of settled beads of Sepharose™ 4 Fast Flow (highly cross linked, 4% agarose derivative). The bead size is approximately 45-165 µm when swollen. Approximately 6.0 mg of Protein A is bound per cc of drained beads. Each cc of drained beads will bind approximately 35 mg Human IgG. The exclusion limit (Mr) of the beads is 3 x 10^7. The maximum linear flow rate is approximately 1300 cm/hr. This product shows long term stability within pH range of 3 to 9 and short term stability within pH range of 2 to 10.
Catalog No. 50-105-9103
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.