Learn More
Applied Biosystems™ 96-Well Septa Dx
Description
This is a 96-well septa designed for use with Applied Biosystems Dx genetic analyzers.
Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in select countries only.
This is a 96-well septa designed for use with the 3500 Dx/3500xL Dx Genetic Analyzer.
Septa and plate base/retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
Note: The 3500 Dx Series Systems and consumables are for distribution and use in select countries only.
Specifications
Specifications
| Compatibility | 96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates |
| Quantity | 20 Pk. |
| Format | 96-well |
| For Use With (Equipment) | 3500 xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 xL Genetic Analyzer, 3500 Genetic Analyzer |
| Product Type | Septa |
| Shipping Condition | Room Temperature |
| Content And Storage | Ambient storage. Each Pk. contains 20 sets. |
For In Vitro Diagnostic Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.