Learn More
Applied Biosystems™ Septa Cathode Buffer Container Dx
Description
This is a septa cathode buffer container intended for use with Applied Biosystems Dx genetic analyzers.
This is a septa cathode buffer container intended for use with the 3500 Dx/3500xL Dx Genetic Analyzer.
Specifications
Specifications
| Quantity | 10 Ea. |
| For Use With (Equipment) | 3500 xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer |
| Product Type | Septa Cathode Buffer Container |
| Shipping Condition | Room Temperature |
| Content And Storage | Each Pk. contains 10 containers. |
For In Vitro Diagnostic Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.