Learn More
Applied Biosystems™ Septa Cathode Buffer Container, for 3500 and SeqStudio™ Flex
Description
These septa are for both compartments of the Cathode Buffer Container and are for use with 3500 and SeqStudio Flex series genetic analyzers.
Specifications
Specifications
| Quantity | 10 Containers |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers |
| Product Type | Septa Cathode Buffer Container |
| Shipping Condition | Room Temperature |
| Content And Storage | This product contains 10 septa. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.