Learn More
STERIS Septihol™ Ethyl WFI Sterile Alcohol Solution
Catalog No. 19415119 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
22 oz.
32 oz.
1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-119 32 oz.
19-415-118 22 oz.
19-415-120 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-415-119 Supplier STERIS Supplier No. 1S1196
Add to Cart
Add to Cart
This item is not returnable. View return policy
A solution of 70% (v/v) reagent-grade alcohol and 30% (v/v) USP WFI designed to remove organic and inorganic residues in aseptic processing areas; double bagged for easy entry into critical environments.
  • Designed to remove organic and inorganic residue from product, and non-product contact surfaces
  • A mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI)
  • Free of stabilizers and non-volatile denaturants for quick and complete evaporation
  • Ready to use, sterile alcohol eliminates in-house production validation and documentation concerns
  • Packages are double bagged for easy entry into controlled environments and aseptic core areas
  • Includes extensive documentation package to meet validation objectives
  • Each lot documented by a Certificate of Analysis including irradiation and sterility results
  • Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
Product Type Ethyl Alcohol
Sterility Sterile
Filtered Filtered
For Use With (Application) Removing Organic and Inorganic Residues in Aseptic Processing Areas
Quantity 12/Cs.
Volume (English) 32 oz.