Learn More
Septin 9 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Specifications
Specifications
| Antigen | Septin 9 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | SEPT9 |
| Gene Accession No. | Q9UHD8 |
| Gene Alias | AF17q25, KIAA0991, MLL septin-like fusion, MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, MSF, MSF1, NAPB, Ov/Br septin, ovarian/breast septin, PNUTL4, SeptD1, septin D1, SINT1 |
| Gene Symbols | SEPT9 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.