Learn More
Applied Biosystems™ SeqScape™ Software v4.1, initial license
Catalog No. A53548
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 license
Catalog No. Quantity
A53548 1 license
1 options
Catalog No. A53548 Supplier Applied Biosystems™ Supplier No. A53548
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.
Initial license for SeqScape Software, a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation.
  • SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation
  • It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities
TRUSTED_SUSTAINABILITY
Specifications
Product Line SeqScape
Software Type Sequencing Data Analysis
Content And Storage The software is provided in the form of a USB stick.
Data Entry CE Sequencing (.ab1 files)
License Initial
Operating System Windows 10
Product Type SeqScape Software v4.0 Initial License
Quantity 1 license
Software Category Supports files from all AB Genetic Analyzers
How can I back up my SeqScape Software data?
Backing up data can help protect you from data loss and/or for sending data to Technical Support for troubleshooting. To export Projects, Project Templates, Reference Data Groups, Libraries, Analysis Defaults, Analysis Protocols, and Display Settings:
1. Go to the Tools menu and select SeqScape Manager.
2. Click on the tab that contains the file(s) you want to back up or export.
3. Select the file(s) that you want to export. Hold down the Shift key while selecting files to select a group of files, or the Control key to select multiple individual files. Once they are selected, click on Export at the bottom of the window.
4. Browse to the location where you want to save the files, and click Export to save them (Note: When selecting multiple files, the File name will appear as the pathway to the location where you are going to save the files.)
If you are performing a data backup, you will want to repeat steps 2-4 for each tab to make sure that all of your work is backed up. Once you have backed them up, save the files to the storage media of your choice (CD, DVD, network drive, flash drive).
Where can I download the Release Notes for SeqScape Software v4.1?
Please download this PDF (http://downloads.thermofisher.com/SeqScape_Software_v4.1/100107873_SeqScape_v4.1_Release_Notes.pdf).
What has been updated in SeqScape Software v4.1?
The primary updates for SeqScape Software v4.1 are the following:
- SeqScape Software v4.1 is supported only on Windows 10 Professional/Enterprise operating system
- SeqScape Software v4.1 supports data generated from SeqStudio Flex Series Genetic Analyzers
- SeqScape Software v4.1 uses KB Basecaller version 1.4.2.6 for basecalling
Can I install SeqScape Software v4.1 on Windows 7 operating system?
No, SeqScape Software v4.1 is supported only on Windows 10 Professional/Enterprise operating system.
Which instrument data does SeqScape Software v4.1 support?
SeqScape Software v4.1 fully supports data generated from the following Applied Biosystems instruments:
- SeqStudio Flex Series Genetic Analyzers
- SeqStudio Genetic Analyzer
- 3500 Genetic Analyzer
- 3500xL Genetic Analyzer
- 3130 Genetic Analyzer
- 3130xl Genetic Analyzer
- 3730 DNA Analyzer
- 3730xl DNA Analyzer
- 310 Genetic Analyzer
Can I install SeqScape Software on Citrix?
No, we do not support any of our capillary electrophoresis (CE) software products in virtual environments.
Do you have a manual of install/uninstall procedures for SeqScape Software v4?
Please download this PDF (https://downloads.thermofisher.com/Instrument_Software/CE/SeqScape/v4.0/100079199_MNL_SEQSCAPE_V4_INSTALLATION.pdf ) and follow the directions for install/uninstall SeqScape Software v4.
How can I import my SeqScape Software data?
If you are upgrading or reinstalling software, you may want to import your data. To do so:
1. Go to the Tools menu and select SeqScape Manager.
2. Click on the tab where you want to import the data (e.g., if you want to import a project, click on the Projects tab).
3. Click on the Import button and select the files you wish to import. Then click Import.
After clicking on Import, a progress bar will appear on the screen. If the file is imported successfully, it will appear in the list for that tab. If the file is not, an error message will appear explaining why the file was not imported. In the example below, the Project name already existed in the database.
Correct the error by changing the Project name or removing the conflicting Project, and reimport the file.
For Research Use Only. Not for use in diagnostic procedures.