Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ SeqStudio™ Sequence Analysis Software

Catalog No. 4443764
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SeqStudio™ Genetic Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
44-437-64 SeqStudio™ Genetic Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-437-64 Supplier Applied Biosystems™ Supplier No. 4443764
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Sequence Analysis Software for SeqStudio™

Sequence Analysis Software for SeqStudio™

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SeqStudio™ Genetic Analyzer
Product Type Sequence Analysis Software
Content And Storage 2 to 8°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.