Learn More
Bioworld SEQUESTRENETM 138 6% IRON
Supplier: Bioworld 306301141
This item has a minimum order quantity of 5 per supplier requirements. SEQUESTRENETM 138, 6% IRON CHELATE: Sequestrene 138 Synonym: Fe-EDDHA; High performance iron chelates for foliar and soil applications; Replacement for FeNa-EDTA; Plant tissue culture tested; Soluble in water. Iron is essential for the f...
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.