Learn More
Bioworld Sequestrene™ 330, 10% Iron Chelate, 250 g
Supplier:  Bioworld 306301153
Encompass_Preferred
Sequestrene™ 330Synonym: Sodium ferric diethylenetriamine pentaacetate; Fe-EDDHAHigh performance iron chelates for foliar and soil applicationsReplacement for FeNa-EDTAPlant tissue culture testedSoluble in waterIron is essential for the formation of chlorophyll, a vital component to ensure plant health. Since iron has low plant mobility, new growth in iron deficient plants often shows iron deficiency. In early stages, leaves have intervenial chlorosis where plant leaves are light green to yellow but the leaf veins remain green. In severe cases, leaves turn white and plants may die. Sequestrene™ 330 is a strong chelate that maintains and protects iron availability in a wide variety of problem soils. Plus, Sequestrene™ has the added flexibility for either foliar or soil applications. Sequestrene™ 330, a 10 percent fully chelated DTPA iron, performs best in a slightly acid to slightly alkaline soils wit a pH of up to 7.5.
Catalog No. 50-254-789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.