Learn More
CPC Scientific H-Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP048A
Encompass
High affinity protein kinase C pseudosubstrate.ONE-LETTER SEQUENCE: RFARKGSLRQKNVMOLECULAR FORMULA: C67H118N26O17MOLECULAR WEIGHT:1559.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [136795-05-6], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: C. House et al., Science, 238, 1726 (1987)
Catalog No. 50-211-0201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.