Learn More
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophobic bead based on crosslinking requirements for the encapsulation layer
Specifications
Specifications
| Concentration | 5% solids (50mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | DNA Sample Prep, Protein Purification |
| Quantity | 15 mL |
| Diameter (Metric) | 1 μm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.