Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic

Catalog No. 09981119 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
15 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-981-119 100 mL
09-981-118 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-981-119 Supplier Cytiva Supplier No. 44152105050350
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) DNA Sample Prep, Protein Purification
Quantity 100 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.