Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles

Catalog No. 09981916 Shop All Cytiva Products
Change view
Click to view available options
Packaging:
1 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
09-981-916 100 mL
09-981-914 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-981-916 Supplier Cytiva Supplier No. 19152104010350
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.

  • Combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics
  • Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY

    Specifications

    Binding Property Amine
    Concentration 10 mg/mL
    Material Magnetite
    Packaging 100 mL
    Diameter (Metric) 1 μm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.