Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles

Catalog No. 45002358 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-358 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-358 Supplier Cytiva Supplier No. 19152104010150
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Reduce undesired protein adsorption. Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles respond twice as fast and feature a non-surfactant, non-protein blocking reagent. They are the ideal choice for molecular biology, nucleic acid isolation, research and clinical diagnostic immunoassays.

Sera-Mag SpeedBeads Amine-Blocked Particles combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics.
  • Non-surfactant, non-protein blocking reagent provides very low non-specific binding of proteins from the sample matrix
  • Fast reaction kinetics increases throughput and precision, and also enables faster movement through viscous solutions
  • Unique cauliflower-like surface provides increased area for binding reactions compared to smooth surface particles
  • Uniform, nominal 1μm diameter provides excellent lot-to-lot reproducibility
Sera-Mag SpeedBeads Respond Twice As Fast
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. Two layers of magnetite are coated onto this core, resulting in faster magnetic response times. The surface is then blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10mg/ml
Content And Storage 4°C
Quantity 5 mL
Diameter (Metric) 1 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.