Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag SpeedBeads™ Streptavidin-Blocked Magnetic Particles
GSA_VA

Catalog No. 09981913 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-981-913 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-981-913 Supplier Cytiva Supplier No. 21152104010350
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Combine high affinity for biotinylated target molecules with very low nonspecific binding.

  • Combine fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin Coating Targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is then blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 mg/mL
    Content And Storage 4°C
    Description Sera-Mag Magnetic Beads
    Material Magnetite
    Packaging 100 mL
    Quantity 100 mL
    Diameter (Metric) 1 μm
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.