Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ SpeedBeads Streptavidin-Blocked Magnetic Particles

Catalog No. 09928216 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-928-216 1 mL
09-928-212 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-928-216 Supplier Cytiva Supplier No. 29729996
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sera-Mag Streptavidin-Blocked Magnetic SpeedBeads combine high affinity for biotinylated target molecules with very low nonspecific binding.

  • Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine high affinity for biotinylated target molecules with very low nonspecific binding.
  • Very high binding capacity and low non-specific binding for biotinylated target molecules on-surfactant due to non-protein blocking reagent
  • Fast reaction kinetics increases throughput and precision, enabling faster movement through viscous solutions
  • Unique cauliflower-like surface structure provides increased surface area for binding reactions compared to smooth-surfaced particles
  • Uniform, nominal 1 μm and 3 μm diameters provide excellent lot-to-lot reproducibility
  • Also available as custom manufactured beads
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mg/mL
Content And Storage 4°C
Contents Magnetic Beads
Color Black
Material Magnetite
Product Type Dye Reagents
Volume (Metric) 1 mL
Diameter (Metric) 3 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.