Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Xtracta Virus/Pathogen Kit

Catalog No. 09920081 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 Extractions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-920-081 1000 Extractions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-920-081 Supplier Cytiva Supplier No. 29514201
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sera-Xtracta Virus/Pathogen Kit for high-throughput total nucleic acid (DNA/RNA) isolation from bacteria and viruses including Adenovirus (Type 14), Influenza A (H3N2) and COVID-19.

  • Scalability: Up to 400 μL of sample
  • Recovery Specifications: Reproducible detection of low viral titer from 10 pfu/mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage All reagents can be stored at ambient temperature (15 to 25°C). For Proteinase K, once reconstituted, store at 2 to 8°C
For Use With (Application) Nucleic acid extraction
Includes 630 mL Binding/Lysis Reagent; 1000 mL Wash Buffer ; 10.4 mL SeraSil-Mag, 400 beads; 10.4 mL SeraSil-Mag, 700 beads; 2 x 110 mg Proteinase K (lyophilized)
Quantity 1000 Extractions
Test Time Can be completed in less than 30 minutes
Form Liquid, Magnetic Bead Based
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.