Learn More
Description
- Human plasma based controls mimic patient samples
- Independent controls meet compliance requirements of regulatory agencies and professional organizations
- Liquid, ready-to-use: no reconstitution or special handling required
- Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
Specifications
Specifications
| Certifications/Compliance | RUO |
| Format | Vial |
| Type | ACCURUN HBV DNA Positive Control |
| Intended Use | Detect Hepatitis B Virus DNA |
| Matrix | Human Serum or Plasma |
| Quantity | 5 x 0.6 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.