Learn More
Description
- Panel helps labs maintain the highest standards for assay validation, regulatory compliance and method development
- Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing reagent, assay/platform performance, sensitivity and specificity to be independently challenged
- Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
- Minimizes down time and accelerates verification of new test methods, reagent systems and platforms
- SeraCare panels are rigorously tested and characterized using the highest quality test methods
- Comprehensive test results provided with each panel; quantified against international standards when available
- Long-term stability and availability
- All SeraCare facilities are ISO 9001 and 13485 certified and comply with cGMP standards
HCV Seroconversion Panel Mfr. No. PHV922-1.2
- HCV genotype 3a
- Undiluted aliquots from plasma units collected from a 22-year old male in the US Midwest in 2008
- HCV genotype 1a
- Undiluted aliquots from plasma units collected from a 30-year old male in the U.S. Northeast in 2008
Specifications
Specifications
| Content And Storage | -65°C to -80°C |
| Format | Genotype 1a Panel |
| Type | Hepatitis C Seroconversion Panel |
| Intended Use | For evaluating HCV test system using well characterized specimens, to provide comprehensive data for comparative analysis |
| Quantity | 6 x 1.2 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.