Learn More
Description
- Maintain standards for assay validation, regulatory compliance, and method development
- Each panel includes comprehensive, comparative data from a variety of methods and commercially available test kits, allowing user to independently challenge reagent, assay, and platform performance, sensitivity and specificity
- Assembled from highly characterized, disease state patient samples, and developed specifically for infectious disease testing, both serology and NAT
- Evaluate, monitor, and troubleshoot test and staff performance
- Minimizes down time and accelerates verification of new test methods, reagent systems, and platforms
- For research use only
Specifications
Specifications
| Content And Storage | -65°C to -80°C |
| Type | Bleed Dates July/August 2001 |
| Intended Use | Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis |
| Quantity | 6 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.