Learn More
Description
- Group of serial bleeds from an individual plasma donor during seroconversion
- Contains set of undiluted plasma samples from single plasma donor
- 1 vial per member, 1.2mL per vial
Cat. No. 21-123-109:
- Four member panel
- Collected over an 11-day period in 2000
- Member 3 is a low titer HIV antigen sample, challenging for antigen and Ag/Ab tests
- Member 4 is a challenging sample for antibody assays
- Four member panel
- Collected over an 16-day period in 2000
- PRB974-02 is a moderate positive HIV RNA sample, challenging for HIV antigen assays
- PRB974-03 is detected at very low signal to cutoff by all HIV Ag/Ab methods tested
- PRB974-04 is a very challenging sample for anti-HIV assays
- Five member panel
- Collected over an 14-day period in 2002
- PRB975-03 is a challenging sample for HIV RNA assays
- Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
- Four member panel
- Collected over an 9-day period in 1999
- Member 2 is a potentially challenging sample for HIV antigen and Ag/Ab tests
- Demonstrates improvement in sensitivity with introduction of “4th generation” HIV Ag/Ab tests
- Four member panel
- Collected over an 15-day period in 2002
- Member 1 is a challenging HIV RNA sample
- Members 3 and 4 are both challenging for some antibody assays
- Seven member panel
- Collected over an 33-day period in 2010
Specifications
Specifications
| Content And Storage | -10°C |
| Type | HIV-1 Seroconversion Panel |
| Intended Use | Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis |
| Quantity | 4 x 1.2 mL |
| Time Range | 9 Days in 1999 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.