Learn More
Seracare Life Sciences Inc Seraseq Lung & Brain CNV Mix, + 6 copies, 20 uL
Supplier:  Seracare Life Sciences Inc 07100415
Encompass
Single sample with three clinically-relevant CNVs (save acquisition and sequencing costs). Develop, optimize, and validate your assay’s ability to detect a range of copy numbers (amplifications). CNV targets precisely quantitated with digital PCR. Single well-characterized GM24385 human genomic DNA as background wild-type material. Manufactured in GMP-compliant, ISO 13485-certified facilities. This specific CNV Mix for lung and brain cancer includes 6 copies.
Catalog No. NC1894276