Learn More
Description
Specifications
Specifications
| Concentration | 20 mg/mL in water (0.05% sodium azide) |
| Content And Storage | Room Temperature |
| Description | Storage Solution: Water + 0.05% sodium azide |
| Isolation Technology | Magnetic Bead |
| Diameter (Metric) | 400 nm |
| For Use With (Application) | Chaotropic salt chemistry and vortex mixing, roller mixing and sonication |
| Product Type | Silica coated, core-shell structure |
| Quantity | 5 mL |
| Size | 400 nm diameter particles |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.